Learn More
Kern Sohn Internal Rechargeable Battery Pack FCB-A01
Rechargeable battery pack for use with bench scales.
Brand: Kern Sohn FCB-A01
This item is not returnable.
View return policy
Description
- Suitable for bench scale KERN FCB (operating time up to 80 h without backlight, charging time approx. 10 h) precision balance KERN 440 (operating time up to 80 h without backlight, charging time approx. 10 h)
Specifications
Bench Scales | |
Rechargeable Battery Pack |