missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dynabeads™ DNA DIRECT™ Blood Kit

The isolated DNA is free of RNA and contaminants that may inhibit PCR. The isolated DNA is of high integrity and high molecular weight, and PCR-results show excellent reproducibility in combination with high level of sensitivity.

Brand:  Invitrogen™ 63102

Product Code. 10738013

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers

Includes: Dynabeads (in Lysis buffer), red cell lysis buffer, washing buffer and resuspension buffer
This item is not returnable. View return policy

Description

Description

The isolated DNA is free of RNA and contaminants that may inhibit PCR. The isolated DNA is of high integrity and high molecular weight, and PCR-results show excellent reproducibility in combination with high level of sensitivity. The process of DNA isolation relies upon cell lysis to release DNA and adsorption of DNA to the surface of Dynabeads. This is followed by magnetic separation of intact DNA/Dynabeads complex and subsequent washing to remove any residual contaminants and potential PCR inhibitors from isolated DNA. Finally, complex is resuspended for direct use in downstream PCR-reactions. Alternatively, DNA may be eluted from Dynabeads following short incubation at 65°C. Multiple samples may be handled in parallel and isolation process can be automated.

  • Cost-effective, simple and efficient single-tube procedure, completed in only 15 minutes
  • Supports 100 isolations from 100 μL of blood per sample
  • Sufficient for at least 100 PCR amplifications
  • Protocol can be modified to allow isolation from 500 μL blood samples, providing enough template DNA for 1,000 PCR amplifications
  • Isolation of PCR-ready genomic DNA directly from blood
  • <100 μL Blood Samples (Small-Scale), 100μL - 1mL Blood Samples ( Medium-Scale)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in refrigerator (2–8°C).
Blood
Not High-throughput Compatible (Manual)
DNA DIRECT™, DYNAL™, Dynabeads™
100 preps
Genomic DNA
Magnetic Bead
Sequencing
< 100 μL Blood Samples (Small-scale), 100 μL to 1 mL Blood Samples (Medium-scale)
Blood Kit
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For research use only. Not for use in diagnostic procedures.