Learn More
Invitrogen™ Gateway™ pDONR™/Zeo Vector
Enables rapid cloning of one or more genes into virtually any protein expression system
Brand: Invitrogen™ 12535035
Includes: 6μg pDONR/Zeo vector, lyophilized in TE buffer, pH 8.0; 1.25mL Zeocin. Zeocin is provided as a 100mg/mL solution in deionized, sterile water.
Description
- Designed to create entry clone following PCR with attB-modified custom primers
- With entry clone, recombine gene of interest into variety of expression vectors adapted for use in Gateway technology
- PCR product is directionally cloned with efficiencies typically >99%
- pDONR/Zeo vector has pUC origin for high plasmid yields and universal M13 sequencing sites for ease of use
Cloning, Gateway Cloning
Specifications
Gateway System Destination Expression Vector | |
Gateway | |
pDONR |
6 μg pDONR™/Zeo vector, lyophilized in TE buffer, pH 8.0. pDONR™/Zeo is also supplied with 1.25 ml Zeocin™. Zeocin™ is provided as a 100 mg/ml solution in deionized, sterile water. Upon receipt, store the pDONR™/Zeo vector at -20°C and the Zeocin™ at -20°C, protected from light. | |
6 μg | |
Gateway™, pDONR™ |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Gateway® pDONR™ Vectors
Version: 250531 REV 29 March 2012
File size: 238.2 kB
Product Content Correction
Product Title
Invitrogen™ Gateway™ pDONR™/Zeo Vector > 6μg