missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ E. coli DNA Ligase

Catalyzes formation of phosphodiester bonds in presence of β-NAD between double-stranded DNAs with 3' hydroxyl and 5' phosphate cohesive termini

Brand:  Invitrogen™ 18052019

Product Code. 10247242

  • 1358.00 SEK / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.


Includes: 10X Reaction Buffer [188mM Tris-HCl (pH 8.3), 906mM KCl, 46mM MgCl2, 37.5mM DTT, 1.5mM λ-NAD, 100mM (NH4 )2 SO4 ]
This item is not returnable. View return policy

Description

Description

Single-stranded nucleic acids are not substrates for this enzyme.

  • Source: Purified from E. coli 594 (Su-) bearing λ lysogen gt4lop-11 lig+ S7
  • Performance and Quality Testing: Endodeoxyribonuclease and 3' and 5' exodeoxyribonuclease assays; ligation efficiency tested
  • Unit Definition: One unit is amount of enzyme required to give 50% ligation of Hind III-digested λ DNA in 30 min. at 16°C in a final volume of 20μL containing a 5' termini concentration of 0.12 'M (300μg/mL)
  • Unit Reaction Conditions: 18.8mM Tris-HCl (pH 8.3), 90.6mM KCl, 4.6mM MgCl2 , 3.8mM DTT, 0.15mM λ-NAD, 10mM (NH4 )2 SO4 in 20μL for 1 hr. at 16°C

Second-strand cDNA synthesis; T4 DNA Ligase alternative when blunt-end ligation is not required; Cloning, Restriction Enzyme Cloning

Order Info

Shipping Conditions: Approved for shipment on wet or dry ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

E. coli DNA Ligase is supplied with a vial of 10X reaction buffer [188 mM Tris-HCl (pH 8.3), 906 mM KCl, 46 mM MgCl2 , 37.5 mM
DTT, 1.5 mM λ-NAD, 100 mM (NH4 )2 SO4 ]. Store at -20°C.
Ligase
100 U
Approved for shipment on Wet or Dry Ice
10X Reaction Buffer
DNA Ligase
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.