Learn More
IRS4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA537926
3023.48 SEK valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
This antibody is predicted to react with rat based on sequence homology.
IRS4 Polyclonal antibody specifically detects IRS4 in Mouse samples. It is validated for Immunocytochemistry, Western Blot.
Specifications
IRS4 | |
Polyclonal | |
Unconjugated | |
IRS4 | |
160 kDa phosphotyrosine protein; Insulin receptor substrate 4; Irs4; IRS-4; Phosphoprotein of 160 kDa; pp160; py160; RP23-415G1.3 | |
Goat | |
Antigen affinity chromatography | |
RUO | |
16370 | |
-20°C | |
Liquid |
Immunocytochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9Z0Y7 | |
IRS4 | |
Peptide with sequence C-DRGTKERKEAKEVRD, from the internal region of mouse Irs4 (aa633-647) | |
100 μg | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Product Title
IRS4 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction