missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ISL1 Monoclonal Antibody (1B1), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA515516
This item is not returnable.
View return policy
Description
MA5-15516 targets ISL1 in indirect ELISA, IF and WB applications and shows reactivity with Human samples. The MA5-15516 immunogen is purified recombinant fragment of human ISL1 expressed in E. Coli. MA5-15516 detects ISL1 which has a predicted molecular weight of approximately 39kDa.
This gene encodes a member of the LIM/homeodomain family of transcription factors. The encoded protein binds to the enhancer region of the insulin gene, among others, and may play an important role in regulating insulin gene expression. The encoded protein is central to the development of pancreatic cell lineages and may also be required for motor neuron generation. Mutations in this gene have been associated with maturity-onset diabetes of the young.Specifications
ISL1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
P61371, P61372 | |
ISL1 | |
Purified recombinant fragment of human ISL1 expressed in E. Coli. | |
RUO | |
16392, 3670 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunocytochemistry, Western Blot | |
1B1 | |
Unconjugated | |
ISL1 | |
Insulin gene enhancer protein ISL-1; insulin related protein; ISL LIM homeobox 1; Isl1; Isl-1; isl-1 homeobox; ISL1 transcription factor LIM/homeodomain (islet-1); ISL1 transcription factor, LIM/homeodomain; ISL1 transcription factor, LIM/homeodomain 1; isl-1=homeobox; Islet 1; ISLET1; islet-1; Lim-homeodomain protein Islet1 | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |