missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isobutylene oxide, 99+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Isobutylene oxide is employed as an initiator for the preparation of polyisobutylenes. It finds application in the synthesis of 1-tert-butylperoxy-2-methyl-propan-2-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Isobutylene oxide is employed as an initiator for the preparation of polyisobutylenes. It finds application in the synthesis of 1-tert-butylperoxy-2-methyl-propan-2-ol.
Solubility
Miscible with ethanol and diethyl ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Isobutylene oxide is employed as an initiator for the preparation of polyisobutylenes. It finds application in the synthesis of 1-tert-butylperoxy-2-methyl-propan-2-ol.
Solubility
Miscible with ethanol and diethyl ether. Slightly miscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
Density | 0.808 |
Boiling Point | 51°C to 52°C |
Flash Point | −32°C (−25°F) |
Odor | Ether-like |
Refractive Index | 1.375 |
Quantity | 25 g |
UN Number | UN2924 |
Beilstein | 102408 |
Solubility Information | Miscible with ethanol and diethyl ether. Slightly miscible with water. |
Formula Weight | 72.11 |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction