Learn More
Isonicotinic acid hydrazide, 99%, Thermo Scientific Chemicals
415.16 SEK - 2030.14 SEK
Chemical Identifiers
CAS | 54-85-3 |
---|---|
Molecular Formula | C6H7N3O |
Molecular Weight (g/mol) | 137.14 |
MDL Number | MFCD00006426 |
InChI Key | QRXWMOHMRWLFEY-UHFFFAOYSA-N |
Synonym | isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide |
PubChem CID | 3767 |
ChEBI | CHEBI:6030 |
IUPAC Name | pyridine-4-carbohydrazide |
SMILES | C1=CN=CC=C1C(=O)NN |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10225120
|
Thermo Scientific Acros
122600050 |
5 g | Glass Bottle |
415.16 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607472
|
Thermo Scientific Acros
122601000 |
100 g | Plastic bottle |
711.69 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10744251
|
Thermo Scientific Acros
122605000 |
500 g | Plastic bottle |
2030.14 SEK
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
54-85-3 | |
137.14 | |
QRXWMOHMRWLFEY-UHFFFAOYSA-N | |
3767 | |
pyridine-4-carbohydrazide |
C6H7N3O | |
MFCD00006426 | |
isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide | |
CHEBI:6030 | |
C1=CN=CC=C1C(=O)NN |
Specifications
54-85-3 | |
Colorless or White | |
Authentic | |
C6H7N3O | |
22, I, 504 | |
isoniazid, isonicotinic acid hydrazide, isonicotinohydrazide, rimifon, isoniazide, nydrazid, isonicotinic hydrazide, laniazid, isonicotinylhydrazine, andrazide | |
QRXWMOHMRWLFEY-UHFFFAOYSA-N | |
pyridine-4-carbohydrazide | |
3767 | |
137.14 | |
Crystalline Powder or Crystals |
171°C to 173°C | |
0.5% max. (1 g, 105°C) | |
99% | |
MFCD00006426 | |
15, 5232 | |
Solubility in water: 14% in water (25 c). Other solubilities: 2% in alcohol (25 c),0.1% in chloroform | |
C1=CN=CC=C1C(=O)NN | |
137.14 | |
CHEBI:6030 | |
99% | |
Isonicotinic acid hydrazide, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 200-214-6
missing translation for 'rtecsNumber' : NS1751850
missing translation for 'tsca' : TSCA
- Isonicotinic acid hydrazide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only