Learn More
Isophthalaldehyde, 98%, Thermo Scientific Chemicals
497.00 SEK - 5535.00 SEK
Chemical Identifiers
CAS | 626-19-7 |
---|---|
Molecular Formula | C8H6O2 |
Molecular Weight (g/mol) | 134.134 |
MDL Number | MFCD00003372 |
InChI Key | IZALUMVGBVKPJD-UHFFFAOYSA-N |
Synonym | isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x |
PubChem CID | 34777 |
IUPAC Name | benzene-1,3-dicarbaldehyde |
SMILES | C1=CC(=CC(=C1)C=O)C=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11482448
|
Thermo Scientific Alfa Aesar
A15788.06 |
5 g |
497.00 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11492448
|
Thermo Scientific Alfa Aesar
A15788.14 |
25 g |
1655.00 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11402458
|
Thermo Scientific Alfa Aesar
A15788.22 |
100 g |
5535.00 SEK
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsophthalaldehyde is used in the synthesis of binuclear ruthenium complex. It participates in base-catalyzed Knoevenagel condensation reaction.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Store at room temperature. Keep away from oxidizing agents.
Chemical Identifiers
626-19-7 | |
134.134 | |
IZALUMVGBVKPJD-UHFFFAOYSA-N | |
34777 | |
C1=CC(=CC(=C1)C=O)C=O |
C8H6O2 | |
MFCD00003372 | |
isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x | |
benzene-1,3-dicarbaldehyde |
Specifications
626-19-7 | |
136°C (17 mmHg) | |
MFCD00003372 | |
Air Sensitive | |
Slightly soluble in water. | |
C1=CC(=CC(=C1)C=O)C=O | |
134.134 | |
134.13 | |
Isophthalaldehyde |
85°C to 91°C | |
C8H6O2 | |
1561038 | |
isophthalaldehyde, m-phthalaldehyde, 1,3-benzenedicarboxaldehyde, 1,3-benzenedialdehyde, isophtaldehydes, isophthaldehyde, isophthaldialdehyde, isophtaldehydes french, isophthalic dicarboxaldehyde, unii-lu162b2n9x | |
IZALUMVGBVKPJD-UHFFFAOYSA-N | |
benzene-1,3-dicarbaldehyde | |
34777 | |
98% |
Safety and Handling
missing translation for 'einecsNumber' : 210-935-8
missing translation for 'rtecsNumber' : NT1981000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.