missing translation for 'onlineSavingsMsg'
Learn More

Isophthalic acid, 99%, Thermo Scientific Chemicals

276.00 SEK - 903.29 SEK

Chemical Identifiers

CAS 121-91-5
Molecular Formula C8H6O4
Molecular Weight (g/mol) 166.13
MDL Number MFCD00002516
InChI Key QQVIHTHCMHWDBS-UHFFFAOYSA-N
Synonym isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
PubChem CID 8496
ChEBI CHEBI:30802
IUPAC Name benzene-1,3-dicarboxylic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C(=O)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10479040
View Documents
Thermo Scientific Acros
122650050
5 g Glass bottle
276.00 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
10277693
View Documents
Thermo Scientific Acros
122655000
500 g Plastic bottle
343.30 SEK
500g
Please to purchase this item. Need a web account? Register with us today!
10755761
View Documents
Thermo Scientific Acros
122650020
2 kg Plastic bottle
903.29 SEK
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

121-91-5
166.13
QQVIHTHCMHWDBS-UHFFFAOYSA-N
8496
benzene-1,3-dicarboxylic acid
C8H6O4
MFCD00002516
isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
CHEBI:30802
C1=CC(=CC(=C1)C(=O)O)C(=O)O

Specifications

121-91-5
White
99%
C6H4(CO2H)2
09, 832
isophthalic acid, 1,3-benzenedicarboxylic acid, m-phthalic acid, m-benzenedicarboxylic acid, acide isophtalique, kyselina isoftalova, iso-phthalic acid, acide isophtalique french, kyselina isoftalova czech, unii-x35216h9fj
QQVIHTHCMHWDBS-UHFFFAOYSA-N
benzene-1,3-dicarboxylic acid
8496
166.13
Crystalline Powder
341.0°C to 343.0°C
Authentic
C8H6O4
MFCD00002516
15, 5243
Solubility in water: 0.01g/100mL (25 c). Other solubilities: freely soluble in alcohol and glacial acetic acid,practically insoluble in benzene and petroleum,ether
C1=CC(=CC(=C1)C(=O)O)C(=O)O
166.13
CHEBI:30802
99%
Isophthalic acid
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 204-506-4

Product Identifier
  • Isophthalic acid

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.