Learn More
Merck Isopore™ Membrane Filters, 0.4 μm
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
1290.00 SEK - 6340.00 SEK
Specifications
Filter Type | Screen Filter |
---|---|
Material | Polycarbonate |
Type | Filter Membranes |
Product Code | Brand | Diameter (Metric) | Color | Pore Size | Bubble Point | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Color | Pore Size | Bubble Point | Price | Quantity & Availability | ||
10557682
|
Merck
HTTP14250 |
142 mm | White | 0.4 μm | 29 psi |
6035.00 SEK
Pack of 50 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
10752381
|
Merck
HTTP01300 |
13 mm | White | 0.4 μm | 29 psi |
1615.00 SEK
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
10687891
|
Merck
HTBP01300 |
13 mm | Black | 0.4 μm | 29 psi |
1605.00 SEK
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
10350181
|
Merck
RTTP01300 |
13 mm | White | 1.2 μm | 8.5 psi |
1605.00 SEK
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
10084500
|
Merck
HTTP02500 |
25 mm | White | 0.4 μm | 29 psi |
1290.00 SEK
Pack of 100 |
Estimated Shipment: 15-08-2024
Log in to see stock. |
||
10074530
|
Merck
HTBP02500 |
25 mm | Black | 0.4 μm | 29 psi |
2860.00 SEK
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
10116570
|
Merck
HTTP03700 |
37 mm | White | 0.4 μm | ≥1.5 bar |
2445.00 SEK
Pack of 100 |
Estimated Shipment: 16-08-2024
Log in to see stock. |
||
10657301
|
Merck
HTTP04700 |
47 mm | White | 0.4 μm | 29 psi |
2850.00 SEK
Pack of 100 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
10339940
|
Merck
HTBP04700 |
47 mm | Black | 0.4 μm | 29 psi |
6340.00 SEK
Pack of 100 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
||
10024330
|
Merck
HTTP09030 |
90 mm | White | 0.4 μm | 29 psi |
2295.00 SEK
Pack of 30 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
10480221
|
Merck
RTTP14250 |
142 mm | White | 1.2 μm | 8.5 psi |
6100.00 SEK
Pack of 50 |
Estimated Shipment: 20-08-2024
Log in to see stock. |
||
Description
Use for analyses of airborne contaminants and other particles using optical or electron microscopy. Isopore™ Polycarbonate Membrane Filters have a smooth, glass-like surface for clearer sample observation. Available in a variety of pore sizes/flow rates, diameters and colors.
The Isopore™ membrane is a polycarbonate, track-etched screen filter recommended for all analyses in which the sample is viewed on the surface of the membrane. Isopore™ membrane offers distinct advantages for the analysis of airborne contaminants and other particles using optical or electron microscopy. The Isopore™ membrane is composed of polycarbonate film, which has a smooth, glass-like surface for clearer sample observation. The unique manufacturing process of the membrane ensures a precise pore diameter and a consistent pore size for accurate separation of samples by size. Matched-weight filters are not usually required because of low, constant tar and ash weights.Features & Benefits:
•Membrane structure retains particles on the surface, simplifying counting and analysis
•Isopore™ membranes do not stain, resulting in low background interference
•Non-hygroscopic, allowing for rapid drying and reduced sample analysis time
•Translucent material does not require clearing for transmitted light microscopy; also available in brown variety
Applications:
Air Monitoring, Epifluorescent Microscopy, Chemotaxis Assays
Specifications
Screen Filter | |
Polycarbonate | |
Filter Membranes |