missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 4-aminobenzoate, 98%, Thermo Scientific Chemicals

Product Code. 11408187
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18144-43-9
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.219
MDL Number MFCD00017108
InChI Key JWCPZKNBPMSYND-UHFFFAOYSA-N
Synonym isopropyl 4-aminobenzoate, 4-amino-benzoic acid isopropyl ester, isopropyl p-aminobenzoate, 4-aminobenzoic acid isopropyl ester, benzoic acid, 4-amino-, 1-methylethyl ester, benzoic acid, p-amino-, isopropyl ester, methylethyl 4-aminobenzoate, isopropyl-4-aminobenzoate, 4-isopropoxycarbonylaniline, acmc-1brt8
PubChem CID 87470
IUPAC Name propan-2-yl 4-aminobenzoate
SMILES CC(C)OC(=O)C1=CC=C(C=C1)N

Specifications

Quantity 100 g
Formula Weight 179.21
Percent Purity 98%
Chemical Name or Material Isopropyl 4-aminobenzoate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'