Learn More
Isopropyl formate, 98%, Thermo Scientific Chemicals
644.00 SEK - 1790.00 SEK
Chemical Identifiers
CAS | 625-55-8 |
---|---|
Molecular Formula | C4H8O2 |
Molecular Weight (g/mol) | 88.11 |
MDL Number | MFCD00014127 |
InChI Key | RMOUBSOVHSONPZ-UHFFFAOYSA-N |
Synonym | isopropyl formate, formic acid, 1-methylethyl ester, formic acid isopropyl ester, formic acid, isopropyl ester, isopropyl methanoate, 1-methylethyl formate, methylethyl formate, unii-h1l164w42g, fema no. 2944, hcooch ch3 2 |
PubChem CID | 12257 |
IUPAC Name | propan-2-yl formate |
SMILES | CC(C)OC=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11453447
|
Thermo Scientific Alfa Aesar
A17222.18 |
50 g |
644.00 SEK
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463447
|
Thermo Scientific Alfa Aesar
A17222.30 |
250 g |
1790.00 SEK
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Isopropyl formate, is a flavoring agent. It can also be used as fragrance agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIsopropyl formate, is a flavoring agent. It can also be used as fragrance agent.
Solubility
slightly soluble in water, completely miscible with alcohol, ether, and most organic solvents.
Notes
d^2^0 0.901. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
625-55-8 | |
88.11 | |
RMOUBSOVHSONPZ-UHFFFAOYSA-N | |
12257 | |
CC(C)OC=O |
C4H8O2 | |
MFCD00014127 | |
isopropyl formate, formic acid, 1-methylethyl ester, formic acid isopropyl ester, formic acid, isopropyl ester, isopropyl methanoate, 1-methylethyl formate, methylethyl formate, unii-h1l164w42g, fema no. 2944, hcooch ch3 2 | |
propan-2-yl formate |
Specifications
625-55-8 | |
0.88 | |
−6°C (21°F) | |
HCO2CH(CH3)2 | |
MFCD00014127 | |
1735844 | |
slightly soluble in water,completely miscible with alcohol,ether,and most organic solvents. | |
CC(C)OC=O | |
88.11 | |
88.11 | |
Isopropyl formate |
-93°C | |
68°C to 71°C | |
C4H8O2 | |
1.3685 | |
UN1281 | |
isopropyl formate, formic acid, 1-methylethyl ester, formic acid isopropyl ester, formic acid, isopropyl ester, isopropyl methanoate, 1-methylethyl formate, methylethyl formate, unii-h1l164w42g, fema no. 2944, hcooch ch3 2 | |
RMOUBSOVHSONPZ-UHFFFAOYSA-N | |
propan-2-yl formate | |
12257 | |
98% |
Safety and Handling
GHS H Statement
H225-H319-H335-H336
Highly flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
May cause drowsiness or dizziness.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H225-H319-H335-H336
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPYL FORMATES
missing translation for 'einecsNumber' : 210-901-2
missing translation for 'rtecsNumber' : LQ8750000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.