Learn More
Isoquinoline-3-carboxylic acid hydrate, 99%, Thermo Scientific Chemicals Product Code.: 11428717
Isoquinoline-3-carboxylic acid hydrate, 99%, Thermo Scientific Chemicals
Product Code. 11428717
5 g
Click to view available options
Quantity:
1 g
5 g
25 g
Product Code. 11428717
Brand: Thermo Scientific Alfa Aesar A19880.06
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 207399-25-5
Molecular Formula C10H7NO2
Molecular Weight (g/mol) 173.17
MDL Number MFCD00075137
InChI Key KVMMIDQDXZOPAB-UHFFFAOYSA-N
Synonym isoquinoline-3-carboxylic acid hydrate, 3-isoquinolinecarboxylic acid hydrate, isoquinoline-3-carboxylic acid monohydrate, isoquinoline-3-carboxylic acid hydrate 1:x, 3-isoquinolinecarboxylic acid, hydrate, 3-isoquinolinecarboxylic acid hydrate 1:1, pubchem10167, acmc-1clhb, 3-isoquinolinecarboxylicacid hydrate, isoquinoline 3-carboxylic acid hydrateShow MoreShow Less
PubChem CID 2736704
SMILES OC(=O)C1=CC2=CC=CC=C2C=N1
Specifications
Melting Point 166°C to 168°C
Quantity 5 g
Beilstein 125548
IUPAC Name isoquinoline-3-carboxylic acid
Formula Weight 173.17 (Anhydrous)
Percent Purity 99%
Chemical Name or Material Isoquinoline-3-carboxylic acid hydrate
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10131074 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberA19880.03,A19880.06
Lot Number10098364 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19880.03,A19880.06
RUO – Research Use Only
Product Content Correction
Product Title
Isoquinoline-3-carboxylic acid hydrate, 99%, Thermo Scientific Chemicals >