Learn More
KGW Isotherm™ Isotherm Borosilicate Glass Insulating Lid
Brand: KGW Isotherm™ 17218
This item is not returnable.
View return policy
Specifications
2000 mL | |
Flexible Applications In Laboratory | |
1 |
Borosilicate Glass | |
Dewar Container; Type 16 Plugs | |
Insulating Lid |
Product Content Correction
Product Title
KGW Isotherm™ Isotherm Borosilicate Glass Insulating Lid > Capacity: 2000mL; For use with: Type 16 Plugs
Spot an opportunity for improvement?Share a Content Correction