missing translation for 'onlineSavingsMsg'
Learn More

ITGA5 Monoclonal Antibody (10F6), Invitrogen™

Mouse Monoclonal Antibody

Brand:  Invitrogen MA515568

Product Code. 11345533

  • 4850.00 SEK / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

MA5-15568 targets ITGA5 in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human samples. The MA5-15568 immunogen is purified recombinant fragment of human ITGA5 expressed in E. Coli. MA5-15568 detects ITGA5 which has a predicted molecular weight of approximately 114kDa.

The product of this gene belongs to the integrin alpha chain family. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This gene encodes the integrin alpha 5 chain. Alpha chain 5 undergoes post-translational cleavage in the extracellular domain to yield disulfide-linked light and heavy chains that join with beta 1 to form a fibronectin receptor. In addition to adhesion, integrins are known to participate in cell-surface mediated signalling.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ITGA5
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
P08648
Itga5
Purified recombinant fragment of human ITGA5 expressed in E. Coli.
RUO
3678
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
10F6
Unconjugated
Itga5
2-phospho-D-glycerate hydro-lyase; alpha enolase like 1; Alpha-enolase; alpha-enolase; c-myc promoter-binding protein-1; CD49 antigen-like family member E; Cd49e; C-myc promoter-binding protein; ENO1; ENO1L1; Enolase 1; enolase 1, (alpha); enolase-alpha; epididymis secretory protein Li 17; fibronectin receptor alpha polypeptide; fibronectin receptor subunit alpha; fibronectin receptor, alpha polypeptide; fibronectin receptor, alpha subunit; FNRA; HEL-S-17; integrin alpha 5; integrin alpha 5 (fibronectin receptor alpha); integrin alpha-5; Integrin alpha-5 heavy chain; Integrin alpha-5 light chain; Integrin alpha-F; integrin subunit alpha 5; integrin, alpha 5 (fibronectin receptor, alpha polypeptide); ITGA5; Itga5_retired; MBP-1; MBPB1; MPB1; MPB-1; MYC promoter-binding protein 1; NNE; Non-neural enolase; phosphopyruvate hydratase; Plasminogen-binding protein; PPH; tau-crystallin; very late activation protein 5, alpha subunit; VLA5; VLA-5; VLA5A
Mouse
100 μL
Primary
Human
Antibody
IgG2a
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.