missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IUPAC pH Standards, Hach Lange™
Click to view available options
Quantity:
500 mL
Description
- pH Standard, IUPAC, pH7.000 at 25 °C, Certified, 500 mL (Radiometer Analytical)
- Shelf life of 48 months from production date
- Features a concentration of 7.000 ± 0.010 pH at 25°C
- Traceable to IUPAC pH scale and NIST
- Total Control of the traceability chain and calculated uncertainties
- The competence and integrity of a laboratory internationally recognized to be accredited for the measurement of pH and conductivity reference materials
Specifications
Quantity | 500 mL |
Color | Colorless |
Odor | Odorless |
pH | 7 |
Solubility | Soluble in water |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'