Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps is a gas collecting tube with O-ring valves on each end.
Brand:  GPE Scientific™ GST/250
 View more versions of this product
Product Code. 15874774
  • 1885.00 SEK / Each
Estimated Shipment: 28-01-2025
to see stock.
Explore more special offers
Description
  • Cylindrical, oblong shape
  • Gas tight container
  • Gauged volume
  • Standard collecting tube with valved ends
Specifications
Specifications
Gas Sample Tube
250 mL