Learn More
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum
GPE Scientific™ J Young Gas Sample Tube with O-Ring Taps and Septum is a gas collecting tube with O-ring valves on each end and a centrally located septum.
1895.00 SEK - 2300.00 SEK
Product Code | Brand | Measurement Range | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Measurement Range | Price | Quantity & Availability | |||||
15874764
|
GPE Scientific™
GSS/100 |
100 mL |
2075.00 SEK
Each |
Estimated Shipment: 30-09-2024
Log in to see stock. |
|||||
15894764
|
GPE Scientific™
GSS/125 |
125 mL |
2085.00 SEK
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||||
15824774
|
GPE Scientific™
GSS/500 |
500 mL |
1940.00 SEK
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||||
15814774
|
GPE Scientific™
GSS/250 |
250 mL |
1895.00 SEK
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||||
15884764
|
GPE Scientific™
GSS/1000 |
1000 mL |
2045.00 SEK
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||||
15804774
|
GPE Scientific™
GSS/2000 |
2000 mL |
2300.00 SEK
Each |
Estimated Shipment: 18-11-2024
Log in to see stock. |
|||||
Description
- Cylindrical, oblong shape
- Gas tight container
- Gauged volume
- Standard collecting tube with valved ends
- Includes centralized septum
Specifications
Gas Sample Tube |