Learn More
GPE Scientific™ J Young O-Rings for Spherical Joints
GPE Scientific™ J Young O-Rings for Spherical Joints facilitate a secure joint seal for spherical joints and are available as Viton or PTFE-coated Viton.
Marke: GPE Scientific™ O40/V
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
- Suitable for use with spherical joints
- Available in Viton or PTFE-coated Viton
- Available in different joint sizes
Spezifikation
Viton | |
O-ring for Spherical Joint | |
40/25 size spherical joints |