Learn More
GPE Scientific™ J Young Schlenk Flask, Cylindrical with Fine Thread Tap and Cone
GPE Scientific™ J Young Schlenk Flask, Cylindrical with Fine Thread Tap and Cone is specially designed for use with vacuum manifolds and Schlenk lines.
Brand: GPE Scientific™ JCC/50
Additional Details : Weight : 0.50000kg
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap and cone
Specifications
50 mL | |
Fine thread tap and cone | |
Size: 19 |
Vacuum manifolds and Schlenk lines | |
Schlenk Flask |