Learn More
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket is specially designed for use with Schlenk lines and vacuum manifolds.
Brand: GPE Scientific™ JRF/100
This item is not returnable.
View return policy
Description
- Solidly constructed for use with vacuum manifolds
- Designed for use with Schlenk lines
- Includes fine thread tap and socket
Specifications
100 mL | |
Fine thread tap and socket | |
Round |
Schlenk lines and vacuum manifolds | |
Size: 24 |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Product Title
GPE Scientific™ J Young Schlenk Flask, Round Bottom with Fine Thread Tap and Socket > Capacity: 100mL
Spot an opportunity for improvement?Share a Content Correction