missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JAZF1 Mouse anti-Human, Clone: PCRP-JAZF1-1C2, Novus Biologicals™
Mouse Monoclonal Antibody
2470.00 SEK - 4910.00 SEK
Specifications
Antigen | JAZF1 |
---|---|
Clone | PCRP-JAZF1-1C2 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18398462
|
Bio-Techne
NBP3-13885-20UG |
20 μg |
2470.00 SEK
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18321714
|
Bio-Techne
NBP3-13885-100UG |
100 μg |
4910.00 SEK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JAZF1 Monoclonal antibody specifically detects JAZF1 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Protein ArraySpecifications
JAZF1 | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin), Peptide Array | |
Unconjugated | |
Mouse | |
DNA Replication Transcription Translation and Splicing | |
10 mM PBS with 0.05% BSA | |
JAZF zinc finger 1, juxtaposed with another zinc finger gene 1, juxtaposed with another zinc finger protein 1, TAK1-interacting protein 27, TIP27ZNF802DKFZp761K2222, Zinc finger protein 802 | |
Recombinant fragment (around aa1-130) of human JAZF1 protein (exact sequence is proprietary) (Uniprot: Q86VZ6) | |
Primary | |
Store at 4°C. |
PCRP-JAZF1-1C2 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunofluorescence 1-2 ug/ml, Immunoprecipitation 1-2 ug/100-500 ug protein, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
Human | |
Q86VZ6 | |
221895 | |
IgG2a | |
Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title