Learn More
Cole-Parmer™ Jenway™ Glass Cuvettes
Brand:  Cole-Parmer™ 83070-42
4968.50 SEK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 11435019
  • 5230.00 SEK / Each
Estimated Shipment: 15-09-2025
to see stock.
Explore more special offers
Specifications
Cuvette
35.0 mL
Glass
100 mm
Product Title
Cole-Parmer™ Jenway™ Glass Cuvettes > Capacity: Up to 35.0mL