Learn More
Tristel JET with Spray Head, 2 x 800 mL
JET is a powerful sporicidal foam for the cleaning and disinfection of environmental surfaces and non-porous materials in laboratories.
Brand: Tristel CCH010201
This item is not returnable.
View return policy
Description
- Compatible with hard, non-porous materials such as laminate, glass, plastic and stainless steel
- Effective against a wide range of microorganisms within a 1-minute contact time
- Destroys DNA and RNA in a matter of seconds
- Complete compliance with EN 14885:2018 and the latest efficacy regulatory requirements
- Each bottle of JET produces 570 doses of sporicidal foam
- Can be used with your choice of spreader, which does not have to be plastic
Specifications
Foaming Disinfectant | |
Spray Head | |
2 Pk. | |
800 mL |
Product Content Correction
Product Title
Tristel JET with Spray Head, 2 x 800 mL > 2 x 800 mL
Spot an opportunity for improvement?Share a Content Correction