missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emma Safety Footwear Jordan (Ruffneck) Safety Shoes
Sneaker with composite toe cap includes a unique rough neck heel design which is highly supportive and ideal for climbing pallets, ladders and getting in and out of truck cabs even when wet. The virtually impenetrable insole protects from sharp objects like nails and shards of metal.
1120.00 SEK
Specifications
Certifications/Compliance | EN ISO 20345:2011 |
---|---|
Closure Type | Laces |
Color | Blue |
Design | Jordan (Ruffneck) |
Liner Material | Hydro-Tec™ Sanitized Silver |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
15949111
|
Emma Safety Footwear
972549-39 |
39 | N/A | ||||||
15959111
|
Emma Safety Footwear
972549-40 |
40 | N/A | ||||||
15969111
|
Emma Safety Footwear
972549-41 |
41 | N/A | ||||||
15979111
|
Emma Safety Footwear
972549-42 |
42 | N/A | ||||||
15989111
|
Emma Safety Footwear
972549-43 |
43 | N/A | ||||||
15999111
|
Emma Safety Footwear
972549-44 |
44 | N/A | ||||||
15909121
|
Emma Safety Footwear
972549-45 |
45 | N/A | ||||||
15919121
|
Emma Safety Footwear
972549-46 |
46 | N/A | ||||||
15929121
|
Emma Safety Footwear
972549-47 |
47 | N/A | ||||||
15939121
|
Emma Safety Footwear
972549-48 |
48 | N/A | ||||||
Description
- S1P-Safety shoe with sole plate for dry work
- Denim/Nubuck construction
- GripForce™ Easy Twist Outsole
- 3M™ Reflective Striping
- ESD certified
- Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
EN ISO 20345:2011 | |
Blue | |
Hydro-Tec™ Sanitized Silver | |
Composite |
Laces | |
Jordan (Ruffneck) | |
Denim, Nubuck |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title