missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VACUUBRAND™ KF DN 25mm Stainless Steel Centering Ring With FPM Sealing
Brand: VACUUBRAND™ 20660125
This item is not returnable.
View return policy
Description
Specifications
Stainless Steel | |
1 Each | |
KF DN 25 mm | |
Centering Ring |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction