Learn More
Lonza™ KGM™ Gold Keratinocyte Growth Basal Medium
Separately sold keratinocyte basal medium for high and low density NHEK-Normal Human Epidermal Keratinocyte cultures.
Brand: Lonza™ 192151
Additional Details : Weight : 1.00000kg
Description
- Keratinocyte basal medium sold separately from combination media kit.
- Can be used with 1 kit of 192152 (keratinocyte cell growth medium SingleQuots).
- Designed to support both clonal growth and high density keratinocyte proliferation.
- Supports superior growth and morphology of keratinocytes.
Specifications
KBM Gold Basal Medium | |
Keratinocyte Cells | |
500 mL Bottle |