missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KIFC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | KIFC1 |
---|---|
Concentration | 0.4 mg/mL |
Applications | Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850365
|
Proteintech
20790-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16840365
|
Proteintech
20790-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody was raised against the C-terminal region of human KIFC1 protein, and recognizes 55-70 kDa of endogenous KIFC1 protein in multiple tissue lysates.
This gene encodes a member of the SWI2/SNF2 family of proteins, which play a critical role in altering protein-DNA interactions. The encoded protein has been shown to have dsDNA-dependent ATPase activity and RNA polymerase II termination activity. This protein interacts with cell division cycle 5-like, associates with human splicing complexes, and plays a role in pre-mRNA splicing.Specifications
KIFC1 | |
Immunocytochemistry, Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q5XI63, Q9BW19, Q9QWT9 | |
100502766, 294286, 3833 | |
KIFC1 Fusion Protein Ag14492 | |
Primary | |
-20°C | |
KIFC1 |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
HSET, KIFC1, kinesin family member C1, Kinesin like protein 2, Kinesin like protein KIFC1, Kinesin related protein HSET, KNSL2 | |
KIFC1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |