missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Kimble™ Aluminum Seal Caps

Product Code. 11822963
Click to view available options
Neck Finish:
11
Closure Liner:
PTFE/Natural Silicone Rubber
PTFE/Rubber
Color:
Natural
Red
No. per Case:
100 Cs.
1000 Cs.
This item is not returnable. View return policy
This item is not returnable. View return policy

With PTFE-faced rubber septa

  • Moderate to good resealability
  • One piece

  • PTFE-faced red rubber septa
  • For general GC applications
  • Moderate resealability
  • Not recommended for multiple injections
  • PTFE-faced natural rubber septa
  • Good resealability
  • Multiple injections

TRUSTED_SUSTAINABILITY

Specifications

Color Natural
For Use With (Application) Multiple injection
Material Aluminum
No. per Case 100 Cs.
Neck Finish 11
Closure Liner PTFE/Natural Silicone Rubber
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.