Learn More
DWK Life Sciences Kimble™ KIMAX™ Accuform™ Micro-Vial™ Vials
Conical cavity allows maximum retrieval of contents with a syringe needle.
Brand:  DWK Life Sciences 60715-310
 View more versions of this product
Product Code. 16438899
  • 2510.00 SEK / Pack of 12
Estimated Shipment: 16-12-2024
to see stock.
Explore more special offers
Description
  • Clear glass Micro-Vial with open top closure and PTFE-faced silicone septum
  • Made with extra thick clear glass walls for both sturdiness and magnification of contents
  • Ungraduated, with marking spot
  • Tube has a screw thread finish
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTFE/Silicone
14 mm
0.3 mL
Tapered
0.08 dr.
36 mm
Amber
Borosilicate Glass
No
Screw
Screw Cap (Open-top)
USP Type I