Learn More
Thermo Scientific™ KingFisher™ Presto Purification System
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
POA - 44710.00 SEK
Product Code | Brand | Description | Samples per Run | Volume (Metric) Processing | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Samples per Run | Volume (Metric) Processing | Price | Quantity & Availability | |||
15881647
|
Thermo Scientific™
5400830 |
KingFisher Presto with 96 DW head | 96 | 50 to 1000 μL |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
15831647
|
Thermo Scientific™
24078830 |
KingFisher Presto 96 Deep Well head | 96 | 50 to 1000 μL |
40090.00 SEK
Each |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15841647
|
Thermo Scientific™
24078831 |
KingFisher Presto 96 Deep Well head with 2 heating blocks | 96 | 50 to 1000 μL |
44710.00 SEK
Each |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15851647
|
Thermo Scientific™
24078840 |
KingFisher Presto 24 Deep Well head | 24 | 200 to 5000 μL |
40060.00 SEK
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15861647
|
Thermo Scientific™
24078841 |
KingFisher Presto 24 DW head with heating block | 24 | 200 to 5000 μL |
43010.00 SEK
Each |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
15891647
|
Thermo Scientific™
5400840 |
KingFisher Presto with 24 DW head | 24 | 200 to 5000 μL |
POA
Each |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||
This item requires a price update prior to being ordered. Request a Price for this item here. |
|||||||||
Description
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories. Utilizing proven KingFisher magnetic particle-based technology, the KingFisher Presto provides high-quality yields of target nucleic acids and proteins for biopharma, biotech and research.
Features:
• Purify 96 or 24 samples at a time
• Volume range of 50–5000 μL
• Minimized size for easy fitting to automation systems
• Heating capability for application requiring elevated temperatures
• Easy connection to several liquid handling instruments
• Flexible system for wide variety of applications