Learn More
Ecolab™ Klercide™ Sporicidal Active Chlorine Unit Dose Concentrate
The active chlorine tablets are double bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation
Brand: Ecolab™ 3082590
5989.75 SEK valid until 2025-05-02
Use promo code "24151" to get your promotional price.
Description
- A sterile highly effective rotational sporicide concentrate, to be diluted 1 tub in 10 litres, for large surfaces.
Specifications
12 Pk. | |
170 g |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
- Warning
Hazard Category
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- EUH031-Contact with acids liberates toxic gas.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Special Offers
-
Multiple Manufacturers
Cleanroom Essentials Extravaganza
Gear Up Your Controlled EnvironmentPromo Code: 24151Expires: 2025-05-02
Your input is important to us. Please complete this form to provide feedback related to the content on this product.