Learn More
Ecolab™ Klerwipe™ Silicone
Sterile low particulate wipes impregnated with silicone and an IPA carrier
Brand:  Ecolab™ 3079440
Product Code. 16287550
  • 10440.00 SEK / Pack of 300
Explore more special offers
Description
Description
  • The blend is 0.2 micron filtered then filled and double bagged in a Grade C cleanroom before being terminally sterilised by gamma irradiation.
  • A sterile ready-to-use maintenance wipe in a resealable pouch, featuring Klertear on outer wraps, for small surfaces.
Specifications
Specifications
Sterile Silicone Pouch Wipe
200 x 200 mm
20 x 15 Wipes