missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLHL8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1980.00 SEK - 4555.00 SEK
Specifications
Antigen | KLHL8 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18313414
|
Bio-Techne
NBP3-04914-20UL |
20 μL |
1980.00 SEK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18323094
|
Bio-Techne
NBP3-04914-100UL |
100 μL |
4555.00 SEK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLHL8 Polyclonal specifically detects KLHL8 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
KLHL8 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
57563 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
kelch-like 8 (Drosophila), kelch-like protein 8, KIAA1378FLJ46304 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 271-620 of human KLHL8 (NP_065854.3). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title