missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ KRTAP10-2 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Brand: Abnova™ H00386679-T02.100uL
Additional Details : Weight : 0.00010kg
Specifications
Western Blot | |
KRTAP10-2 293T Cell Transient Overexpression Lysate(Denatured) | |
Theoretical M.W. 28.05kDa | |
Transient overexpression cell lysate was tested with anti-KRTAP10-2 antibody (Mfr. No. H00386679-B01) by Western Blot. | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
293T | |
BC146565 | |
pCMV-KRTAP10-2 full-length | |
100 μL | |
Human |