missing translation for 'onlineSavingsMsg'
Learn More

KSHV ORF62 Sheep anti-Virus, Polyclonal, Novus Biologicals™

Sheep Polyclonal Antibody

2490.00 SEK - 4640.00 SEK

Specifications

Antigen KSHV ORF62
Dilution Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18345673
View Documents
Bio-Techne
NBP3-11885-25UG
25 μg
2490.00 SEK
25µg
Please to purchase this item. Need a web account? Register with us today!
18331962
View Documents
Bio-Techne
NBP3-11885-250UG
250 μg
4640.00 SEK
250µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KSHV ORF62 Polyclonal antibody specifically detects KSHV ORF62 in Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot
Specifications

Specifications

KSHV ORF62
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot
Unconjugated
Sheep
Virus
Kaposi's Sarcoma Virus, Kaposi's sarcoma-associated herpesvirus, ORF62
A fusion protein that contains at its amino terminus the maltose binding protein and at its carboxy terminus the protein encoded by the open reading frame 62 (ORF62) (Uniprot: Q2HR66)
Primary
Store at -20C. Avoid freeze-thaw cycles.
Western Blot 1:500, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Dot Blot
Polyclonal
Purified
RUO
PBS
4961461
IgG
Ammonium sulfate precipitation
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.