Learn More
L-Arginine, 99%, MP Biomedicals™ Product Code.: 11313929
L-Arginine, 99%, MP Biomedicals™
Product Code. 11313929 Shop All MP Biomedical Products
500 g
Click to view available options
Quantity:
100 g
500 g
1 kg
Product Code. 11313929
Brand: MP Biomedicals 0210073690
Chemical Identifiers
CAS 74-79-3
Molecular Formula C6H14N4O2
Molecular Weight (g/mol) 174.20
MDL Number MFCD00002635
InChI Key ODKSFYDXXFIFQN-UHFFFAOYNA-N
Synonym l-arginine, arginine, l-+-arginine, l-arg, l +-arginine, s-2-amino-5-guanidinopentanoic acid, h-arg-oh, arginina, arginine, l
PubChem CID 6322
ChEBI CHEBI:16467
SMILES NC(CCCN=C(N)N)C(O)=O
Specifications
Color White
Assay Percent Range 99%
IUPAC Name 2-amino-5-[(diaminomethylidene)amino]pentanoic acid
Quantity 500 g
Formula Weight 174.2
Physical Form Powder
Chemical Name or Material L-Arginine free base
Product Content Correction
Product Title
L-Arginine, 99%, MP Biomedicals™ >