missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L-Aspartic acid monosodium salt monohydrate, 99%
Click to view available options
Quantity:
100 g
500 g
Description
L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.
Solubility
Soluble in cold water.
Notes
Incompatible with strong oxidizing agents.
L-Aspartic acid monosodium salt monohydrate is used as a principal neurotransmitter for fast synaptic excitation.
Solubility
Soluble in cold water.
Notes
Incompatible with strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Solubility Information | Soluble in cold water. |
IUPAC Name | sodium 2-amino-3-carboxypropanoate hydrate |
Formula Weight | 173.11 (155.10 Anhydrous) |
Percent Purity | 99% |
Quantity | 100 g |
Chemical Name or Material | L-Aspartic acid monosodium salt monohydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'