missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cysteic acid monohydrate, 99%, Thermo Scientific Chemicals
1220.36 SEK - 3431.71 SEK
Chemical Identifiers
CAS | 23537-25-9 |
---|---|
Molecular Formula | C3H6NO5S |
Molecular Weight (g/mol) | 168.14 |
MDL Number | MFCD00149544 |
InChI Key | XVOYSCVBGLVSOL-REOHCLBHSA-M |
Synonym | l-cysteic acid monohydrate, unii-vn8x20t18r, cysteic acid monohydrate, l, r-2-amino-3-sulfopropanoic acid hydrate, l-alanine, 3-sulfo-, monohydrate, cysteinesulfonic acid hydrate, h-cys o 2-oh.h2o, l-cysteic acid hydrate, l-cysteic acid monohydrate mi, alanine, 3-sulfo-, monohydrate, l |
PubChem CID | 12308854 |
IUPAC Name | (2R)-2-amino-3-sulfopropanoic acid;hydrate |
SMILES | [NH3+][C@@H](CS([O-])(=O)=O)C([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15216038
|
Thermo Scientific Acros
452840100 |
10 g |
1220.36 SEK
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15226038
|
Thermo Scientific Acros
452840500 |
50 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
23537-25-9 | |
168.14 | |
XVOYSCVBGLVSOL-REOHCLBHSA-M | |
12308854 | |
[NH3+][C@@H](CS([O-])(=O)=O)C([O-])=O |
C3H6NO5S | |
MFCD00149544 | |
l-cysteic acid monohydrate, unii-vn8x20t18r, cysteic acid monohydrate, l, r-2-amino-3-sulfopropanoic acid hydrate, l-alanine, 3-sulfo-, monohydrate, cysteinesulfonic acid hydrate, h-cys o 2-oh.h2o, l-cysteic acid hydrate, l-cysteic acid monohydrate mi, alanine, 3-sulfo-, monohydrate, l | |
(2R)-2-amino-3-sulfopropanoic acid;hydrate |
Specifications
23537-25-9 | |
Authentic | |
C3H6NO5S | |
MFCD00149544 | |
+ 8.00 | |
Solubility in water: soluble. Other solubilities: insoluble in alcohol | |
[NH3+][C@@H](CS([O-])(=O)=O)C([O-])=O | |
168.14 | |
187.17 | |
L-Cysteic acid monohydrate |
253.0°C to 254.5°C | |
98.5% min | |
HO3SCH2CH(NH2)CO2H·H2O | |
+ 8.00 (20.00°C c=5, H2O) | |
l-cysteic acid monohydrate, unii-vn8x20t18r, cysteic acid monohydrate, l, r-2-amino-3-sulfopropanoic acid hydrate, l-alanine, 3-sulfo-, monohydrate, cysteinesulfonic acid hydrate, h-cys o 2-oh.h2o, l-cysteic acid hydrate, l-cysteic acid monohydrate mi, alanine, 3-sulfo-, monohydrate, l | |
XVOYSCVBGLVSOL-REOHCLBHSA-M | |
(2R)-2-amino-3-sulfopropanoic acid;hydrate | |
12308854 | |
99% |
Safety and Handling
Product Identifier
- L-Cysteic acid monohydrate, 99%
RUO – Research Use Only