missing translation for 'onlineSavingsMsg'
Learn More

L-Cysteine [Free Base], MP Biomedicals

Product Code. 11405122
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Formula Weight 121.2
Grade Cell Culture Reagent
Quantity 100 g
Physical Form Crystalline
Chemical Name or Material L-Cysteine [Free Base]
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'