missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Lysine, 99.6%, MP Biomedicals™
535.30 SEK - 1372.70 SEK
Chemical Identifiers
CAS | 39665-12-8 |
---|---|
Molecular Formula | C6H16N2O3 |
Molecular Weight (g/mol) | 164.21 |
MDL Number | MFCD00151035 |
InChI Key | HZRUTVAFDWTKGD-UHFFFAOYNA-N |
Synonym | l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1 |
PubChem CID | 16211825 |
IUPAC Name | 2,6-diaminohexanoic acid hydrate |
SMILES | O.NCCCCC(N)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11441681
|
MP Biomedicals
0219022405 |
5 g |
535.30 SEK
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11451681
|
MP Biomedicals
0219022425 |
25 g |
1372.70 SEK
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
39665-12-8 | |
164.21 | |
HZRUTVAFDWTKGD-UHFFFAOYNA-N | |
16211825 | |
O.NCCCCC(N)C(O)=O |
C6H16N2O3 | |
MFCD00151035 | |
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1 | |
2,6-diaminohexanoic acid hydrate |
Specifications
39665-12-8 | |
C6H16N2O3 | |
l-lysine monohydrate, l-lysine hydrate, lysine monohydrate, l +-lysine monohydrate, l-lysine, hydrate, h-lys-oh h2o, lysine, hydrate, lysine hydrate, l-lysine, monohydrate, l-lysine, hydrate 1:1 | |
O.NCCCCC(N)C(O)=O | |
164.21 | |
164.2 | |
L-lysine, Free Base, Monohydrate |
White | |
MFCD00151035 | |
HZRUTVAFDWTKGD-UHFFFAOYNA-N | |
2,6-diaminohexanoic acid hydrate | |
16211825 | |
Crystalline Powder |