missing translation for 'onlineSavingsMsg'
Learn More

L-Ornithine Hydrochloride, 99%, MP Biomedicals

Product Code. 11413100
Click to view available options
Quantity:
25 g
100 g
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

L-Ornithine is a product of arginase degradation and plays an important role in ammonia metabolism via the urea cycle

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3184-13-2
Molecular Formula C5H13ClN2O2
Molecular Weight (g/mol) 168.621
MDL Number MFCD00064562
InChI Key GGTYBZJRPHEQDG-WCCKRBBISA-N
Synonym l-ornithine hydrochloride, l-ornithine monohydrochloride, h-orn-oh.hcl, l-ornithine hcl, l +-ornithine hydrochloride, l-ornithine, monohydrochloride, s-2,5-diaminopentanoic acid hydrochloride, ornithine hydrochloride van, unii-hbk84k66xh, ornithine l monochlorohydrate
PubChem CID 76654
IUPAC Name (2S)-2,5-diaminopentanoic acid;hydrochloride
SMILES C(CC(C(=O)O)N)CN.Cl

Specifications

Quantity 5 kg
Formula Weight 168.6
Grade Cell Culture Reagent
Physical Form Crystalline
Chemical Name or Material L-Ornithine, 99%, Hydrochloride, This material is essentially free of citrulline and ammonia.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.