missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Ribose, 99%, Thermo Scientific Chemicals

414.01 SEK - 3102.24 SEK

Chemical Identifiers

CAS 24259-59-4
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
MDL Number MFCD00167010
InChI Key HMFHBZSHGGEWLO-ZHZWXSSZNA-N
Synonym l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
PubChem CID 90428
ChEBI CHEBI:47015
IUPAC Name (2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES OC[C@@H]1OC(O)[C@@H](O)[C@H]1O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15550524
View Documents
Thermo Scientific Acros
257910010
1 g
414.01 SEK
1g
Please to purchase this item. Need a web account? Register with us today!
15560524
View Documents
Thermo Scientific Acros
257910050
5 g
1140.52 SEK
5g
Please to purchase this item. Need a web account? Register with us today!
15570524
View Documents
Thermo Scientific Acros
257910250
25 g
3102.24 SEK
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24259-59-4
150.13
HMFHBZSHGGEWLO-ZHZWXSSZNA-N
90428
(2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
C5H10O5
MFCD00167010
l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
CHEBI:47015
OC[C@@H]1OC(O)[C@@H](O)[C@H]1O

Specifications

24259-59-4
Authentic
Glass Bottle
MFCD00167010
+ 20.00 (20.00°C c=2,water)
l-ribose, l-+-ribose, 2s,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-ribose, aldehydo-l-ribo-pentose, ror, l-ribose 9ci, ksc202k1p
HMFHBZSHGGEWLO-ZHZWXSSZNA-N
(2S,3S,4S)-2,3,4,5-tetrahydroxypentanal
90428
150.13
L(+)-Ribose
84.0°C to 86.0°C
98.5% min. (ELSD) (HPLC)
C5H10O5
01,859
+ 20.00
(10% in water) Clear colorless
OC[C@@H]1OC(O)[C@@H](O)[C@H]1O
150.13
CHEBI:47015
99%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 246-110-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only