missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Tyrosine Disodium Salt, MP Biomedicals
Click to view available options
Quantity:
25 g
100 g
Specifications
Specifications
Formula Weight | 225.2 |
Quantity | 25 g |
Chemical Name or Material | L-Tyrosine Disodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'