missing translation for 'onlineSavingsMsg'
Learn More

L-Tyrosine Disodium Salt, MP Biomedicals

Product Code. 11451942
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 122666-87-9
Molecular Formula C9H13NNa2O5
Molecular Weight (g/mol) 261.19
MDL Number MFCD03840558
InChI Key WFTCFVUQORELJZ-CMIWSEPMNA-L
Synonym tyrosine disodium dihydrate, unii-5rfd27dq22, l-tyrosine disodium salt dihydrate, l-tyrosine, sodium salt, hydrate 1:2:2, c9h9no3.2na.2h2o, disodium l-tyrosinate dihydrate
PubChem CID 71587456
IUPAC Name disodium (2S)-2-amino-3-(4-oxidophenyl)propanoate dihydrate
SMILES O.O.[Na+].[Na+].N[C@@H](CC1=CC=C([O-])C=C1)C([O-])=O

Specifications

Formula Weight 225.2
Quantity 100 g
Chemical Name or Material L-Tyrosine Disodium Salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'