Learn More
Labconco™ Exterior Glove Port Covers
May be placed over glove ports with or without gloves attached to facilitate purge/fill operations and preserve inert atmosphere
Brand: Labconco™ 5242200
1797.74 SEK valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.80000kg
Includes: Two clamps
Description
May be placed over glove ports with or without gloves attached to facilitate purge/fill operations and preserve inert atmosphere
- Molded neoprene construction
- For use with all glove boxes
Specifications
3.5 lb. | |
1.6 Kg | |
Enclosure Kit | |
All Glove Box |