Learn More
Labconco™ Hard Duct Exhaust Connector for Precise™ Glove Boxes
Accessories for use with Precise Glove Boxes
Brand: Labconco™ 5230400
This item is not returnable.
View return policy
Description
Epoxy-coated steel, 6 in. diameter, for connecting the glove box to ductwork attached to a remote blower. Remote blower must be balanced with glove box's built-in blower. Requires ductwork and remote blower. For use with Precise HEPA-Filtered Glove Boxes and XPert Weigh Boxes.Specifications
Precise™ HEPA Filtered Glove Box, XPert™ Weigh Box | |
2.3 Kg | |
Connecting the glove box to a remote blower, when by-passing the glove box blower |
5 lb. | |
Hard Duct Exhaust Connector |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Labconco™ Hard Duct Exhaust Connector for Precise™ Glove Boxes > Hard duct exhaust connector; Dia.: 6 in.
Spot an opportunity for improvement?Share a Content Correction