Learn More
Labplas Twirl'EM™ Sterile Sampling Bags, SafetyTabs
Provide a sterile, secure, contaminant-free pliant container that ensures dependable analysis results. Labplas Twirl'EM™ Sterile Sampling Bags with safety tabs incorporate easy-to-pull tabs that open the bag without risk of contaminating the bag interior. They are useful in a variety of sampling applications, including environmental and surface sampling.
4425.00 SEK
Product Code | Brand | Capacity (Metric) | Closure Type | Description | Thickness (English) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Type | Description | Thickness (English) | Price | Quantity & Availability | ||
16235272
|
Labplas
EPL47012 |
1650 mL | 2 Round Wires | Clear body | 4 mil |
4425.00 SEK
Pack of 1000 |
Estimated Shipment: 28-02-2025
Log in to see stock. |
||
Description
- Virgin polyethylene tubing
- Can be heated to 240°C for sterility
- Large bag mouth opening to facilite insertion of samples
- Easy pull tabs to open and close the bag with decreased risk of contamination
- R-Nase, D-Nase, Pyrogen, and BPA free
- Environmental sampling
- Surface sampling
- Carcass sampling
- Biomedical research
- Pharmaceutical research
- Food industry
Certifications
Sterility
Compliance
EPA, FDA, USDA, HACCP
Specifications
Polyethylene |