Learn More
Labplas Twirl'EM™ Sterile Sampling Bags, SafetyTabs
Provide a sterile, secure, contaminant-free pliant container that ensures dependable analysis results. Labplas Twirl'EM™ Sterile Sampling Bags with safety tabs incorporate easy-to-pull tabs that open the bag without risk of contaminating the bag interior. They are useful in a variety of sampling applications, including environmental and surface sampling.
4425.00 SEK
Specifications
Material Polyethylene
View More Specs
Products 1
Product Code Brand Capacity (Metric) Closure Type Description Thickness (English) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Closure Type Description Thickness (English) Price Quantity & Availability  
16235272
View Documents
Labplas
EPL47012
1650 mL 2 Round Wires Clear body 4 mil
4425.00 SEK
Pack of 1000
Estimated Shipment: 28-02-2025
to see stock.
Add to basket
Description
Description
  • Virgin polyethylene tubing
  • Can be heated to 240°C for sterility
  • Large bag mouth opening to facilite insertion of samples
  • Easy pull tabs to open and close the bag with decreased risk of contamination
  • R-Nase, D-Nase, Pyrogen, and BPA free
Applications include:
  • Environmental sampling
  • Surface sampling
  • Carcass sampling
  • Biomedical research
  • Pharmaceutical research
  • Food industry
Certifications
Sterility
Compliance
EPA, FDA, USDA, HACCP
Specifications
Specifications
Polyethylene