Learn More
Lacidipine, MedChemExpress
MedChemExpress Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.
Brand: Medchem Express HY-B0347
This item is not returnable.
View return policy
Description
Lacidipine (Lacipil, Motens) is a L-type calcium channel blocker.Specifications
Lacidipine | |
Off-White | |
C26H33NO6 | |
DMSO : ≥ 50 mg/mL (109.76 mM) | |
455.54 | |
99.98% | |
Solid |
103890-78-4 | |
Research | |
10 mg | |
O=C(C1=C(C)NC(C)=C(C(OCC)=O)C1C2=CC=CC=C2/C=C/C(OC(C)(C)C)=O)OCC | |
455.54 | |
Research | |
COVID-19-immunoregulation |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Lacidipine, MedChemExpress